Thermo Fisher Scientific
Why does one system not want to start?
Environment
AS-AP Autosampler 
Dual IC System 
Resolution
Please ensure that the Release Exclusive Access command is moved up from the end of the method script to about the 3:00 minute timestamp for both channels.