65000-98007 - Rev A - Prelude LX-4 MD Operator Manual
Description:
The Thermo Scientific™ Prelude LX-4 MD™ is a liquid chromatography (LC) instrument that runs high-performance liquid chromatography (HPLC) and ultra-high-performance liquid chromatography (UHPLC). You can run up to four LC channels at the same time by using one detector.
The Prelude LX-4 MD control application controls the Prelude LX-4 MD instrument components, while interfacing with the data processing software. This manual describes how to use the Prelude LX-4 MD control application to manage instrument components and create the LC and autosampler (AS) methods.
Environment:
Thermo Scientific™ Prelude LX-4 MD™
Attachment(s):
| File | Last Modified |
|---|---|
| 65000-98007 - Rev A - Prelude LX-4 MD Operator Manual | May 25, 2022 |