Thermo Fisher Scientific
Error Code 89 - Liquid leak sensor missing or defective
Issue
Vanquish pump experiencing error Code 89: Liquid leak sensor missing or defective
Environment
  • Vanquish Core
    • VC-A12-A-02
    • VC-A13-A-02
  • Vanquish Flex
    • VF-A10-A-02
    • VF-A40-A-02
  • Vanquish Horizon
    • VH-A10-A-02
    • VH-A40-A-02
  • Chromeleon
  • Vanquish Pump
Resolution
  1. Disable the leak sensor functionality in the Chromeleon software by setting Leak Sensor Mode to Disabled.
  2. Contact Thermo Fisher Scientific Technical Support for assistance.
Cause
Hardware failure.