70-4685 - Rev E - Model 5600A CoulArray Detector Operator Manual
Description:
The ESA CoulArray® Detector is a coulometric multi-electrode electrochemical detector for high performance liquid chromatography (HPLC).
The instrument is designed to detect and quantitate trace levels of electroactive compounds in complex matrices. A typical application is the qualitative and quantitative analysis of a broad range of biogenic amines in biological tissues.
The attached document describes the operation of the instrument.
Environment:
Model 5600A CoulArray Detector
Attachment(s):
| File | Last Modified |
|---|---|
| 70-4685 - Rev E - Model 5600A CoulArray Detector Operator Manual.pdf | May 25, 2022 |