Thermo Fisher Scientific
VQ-Packing-Instructions-DOC4801-0026-5-0
Description: 
To prevent the devices of the Thermo Scientific Vanquish series from being damaged during transit when they need to be returned to the factory, always do the following:
• Use the original shipping container or the shipping container mentioned in attached document.
• Observe the packing instructions in attached document.
Inappropriate packaging automatically voids the product warranty. If the original packaging is no longer available, appropriate shipping containers and packing material can be ordered from the Thermo Fisher Scientific sales organization for HPLC products
Environment:
Vanquish systems
  • Was this article helpful?