Vanquish Neo System Installation Qualification Operating Instructions
Description:
These Vanquish Neo System Installation Qualification operating instructions describe the procedures executed during the installation to confirm:
- That the principle communication between SII for Xcalibur / ChromeleonTM and the connected modules is successfully established. It is not intended to completely validate all module’s communication and functions.
- The Vanquish Neo System (or with column compartment) basic chromatographic performance meets installation qualification (IQ) specifications.
The document does not describe a procedure for Operational Qualification of the connected modules.
Environment:
Vanquish Neo
Attachment(s):
| File | Last Modified |
|---|---|
| VanquishNeoSystem_IQReport_V3.xlsm | September 19, 2024 |
| OI-VanquishNeoSystem-Instruments-IQ-3-0.pdf | September 19, 2024 |