Thermo Fisher Scientific
Abnormal liquid detection errors
Issue
Abnormal liquid detection errors in Gallery and Gallery Plus analyzers that make the instrument to stop and go to the idle mode abnormally.
Environment
  • Gallery
  • Gallery Plus
  • Beermaster Analyzer 6.0
  • Beermaster Analyzer 7.0
Resolution
Downgrade the software to the earlier software version or replace the MOTOR4 PCB dispenser MOTOR4 (position 8 in Gallery and position 8 or 10 in Gallery Plus) to another MOTOR4 PCB with a processor revision D.
  • Was this article helpful?