Thermo Fisher Scientific
Checking Reagent Barcodes
Issue
Checking reagent barcodes
Environment
  • Gallery/Gallery Plus/Beermaster Analyzer 7.0
  • Gallery/Gallery Plus/Beermaster Analyzer 6.0
Resolution
To check the content of the reagent rack:
  1. Remove the rack from the analyzer. 
  2. Ensure the barcodes are properly aligned. 
  3. Place the rack back on the analyzer. 
The reagents' barcodes are read and the view is updated.  
  • Was this article helpful?