Thermo Fisher Scientific
Clearing Rack Information
Issue
Clearing rack information
Environment
  • Gallery/Gallery Plus/Beermaster Analyzer 7.0
  • Gallery/Gallery Plus/Beermaster Analyzer 6.0
Resolution
To clear the information from all positions from a rack:
NOTE: The rack has to be outside of the analyzer when the rack information is handled. 
  1. Select F3 > REAGENT RACKS. 
  2. Select the rack from the racks field. 
  3. Select the rack position.
  4. Click 'clear position' to remove the rack position information.