Thermo Fisher Scientific
Daily Maintenance of the Analyzer for Use of Corrosive Reagents
Issue
To reduce the risk of corrosion, special attention should be paid for the daily maintenance of the analyzer. 
Environment
  • Gallery/Gallery Plus/Beermaster Analyzer 7.0
  • Gallery/Gallery Plus/Beermaster Analyzer 6.0
Resolution
  • Empty the cuvette waste bin of used cuvettes preferably after each test run or at least at the end of the day while the analyzer is still on.
  • Flush the cuvette waste bin with water and remove any residue.
  • At the end of the day, take the reagents away from the analyzer. 
  • Open the analyzer's main vent to ventilate the analyzer and leave the cover open for at least 30 minutes.
  • Wipe all visible reagent spills with a cloth moistened with mild detergent. 
  • Empty the waste water container and clean with tap water. 
BE ADVISED: Ensure that the laboratory is adequately and properly ventilated. 
  • Was this article helpful?