How to remove a reagent rack
Issue
How do I remove a reagent rack?
Environment
- Gallery
- Gallery Plus
- Beermaster Analyzer 6.0
- Beermaster Analyzer 7.0
Resolution
To remove a rack reagent:
- Select F3 > REAGENT
- Select the rack to be removed.
- Click 'remove rack.'
- When the light is on, open the cover.
- Remove the rack.
- Close the cover.