Can't Click Get Ready Button
Issue
Qtegra froze and Get Ready button not working
Environment
- iCAP 7000 series
- iCAP 6000 series
- iCAP Q/RQ/TQ
- Qtegra
- Windows 10
- Windows 7
- ECCN: EAR99
Resolution
Restart the Qtegra Services. This knowledge article can be used as a reference on how to do so: Restarting Qtegra Services