Thermo Fisher Scientific
Firmware update failed
Issue
When upgrading to Qtegra 2.14 SR1, 2.14 SR2 or 2.18, a Firmware update was required but the update has failed
Environment
  • iCAP Q
  • iCAP RQ
  • iCAP TQ
  • ECCN: EAR99
Resolution
Qtegra must be first licensed and computer restarted in order for firmware update to successfully take place
Cause
It appears that because the Scan Control firmware is being updated that the software must be licensed in order for this to take place 
  • Was this article helpful?