Loss of some counts
Issue
Loss of some but not all counts
Environment
- iCAP RQ
Resolution
Switched to a different configuration, and ran real time display.
Counts came back in the different configuration.
Restored the original configuration,and was able to see all of the counts again.