Thermo Fisher Scientific
Mass Calibration Verification Failure
Issue
Mass Calibration Verification Failure for Peak Width or Offset in Performance Report
Environment
  • iCAP Q
  • iCAP RQ
  • ECCN: EAR99
Resolution
Perform a mass calibration using the Mass Calibration wizard in the Qtegra software. The mass calibration will need to be performed in STD (or STDS mode if using High Sensitivity interface) using the Setup or Calibration (i.e.P/N Thermo-5A) solution.
  • Was this article helpful?