Plasma switches off
Issue
Plasma goes out before going into operate state directly after the slide valve has opened
Environment
- iCAP Q
- iCAP RQ
- Qtegra
- ECCN: EAR99
Resolution
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Analysis
There could be other parameters, such as lens voltages, which could have changed but would not have caused the plasma to go out. It is a good idea once in Operate state to go to the Edit window (see step 1) and check the other parameters to make sure they match up with the values which were used the last time you successfully ran the instrument. Another way to do this would be to compare the current values in the Control Panel once in Operate state with the values from the most recent passing performance report.
Cause
Most of the time this is due to a plasma related setting being set to ‘0’ in the measurement mode because upon going into Operate state, Qtegra sets the instrument settings to whatever is listed in the measurement mode.