Poor Signal Stability
Issue
Poor signal stability
Environment
- iCAP Q
- iCAP RQ
- iCAP TQ
- ECCN: EAR99
Resolution
To troubleshoot signal stability issues follow these steps:
- Check the signal stability in all measurement modes.
- If poor signal stability can be observed in all modes, proceed with step 2.
- If poor signal stability occurs in modes using the QCell pressurized with gas only, proceed with step 11.
- Make sure that the total dissolved solids level of the solution being aspirated is below the recommended levels.
- Make sure that the laboratory temperature is stable.
- Make sure that the temperature of the cooling water supplied to the instrument is stable.
- Make sure that the gas lines are securely connected, free from kinks, and not trapped.
- Make sure that the gas flow readback values in the Dashboard are stable.
- Make sure that the torch body is aligned correctly inside the torch holder.
- Make sure that the waste tubing is inserted in the waste vessel and above the waste level.
- Make sure that the pump speed of the peristaltic pump is stable.
- Make sure that the pressure in the vacuum system is stable.
- Check that all CCT/CR gas connections are okay.
- Run the system for approximately 30 minutes with gas flows set to fully purge and equilibrate the system.
- Run an autotune sequence.
Cause
- Sample matrix can affect stability; especially if the nebulizer cannot form a stable aerosol.
- System-related stability problems may occur if the % RSD values in the short term stability test in the Performance Report are higher than 2% when running a standard tune solution.