Thermo Fisher Scientific
Loss of Emission (LOE) - spontaneous emission loss from the LMIS
Issue
Loss of Emission (LOE) - spontaneous emission loss from the LMIS
Environment
  • Phoenix
Resolution
These are possible resolutions for the issue:
  • Read SB-2020-049 for more details. Use the criteria in this Service Bulletin to determine if 'Excessive LOEs' are occurring (identified as having LOEs with less than 50 hours between each LOE during a 300 hour time period).
Cause
These are possible causes for the issue:
  • Leakage currents, or intermittent sporadic leakage currents, interfere with Steadfast operation and the regulation of LMIS emission