Thermo Fisher Scientific
Cassette gripper and Docker temperature rising during and after the cassette docking and undocking
Issue
Cassette gripper and Docker temperature rising during and after the cassette docking and undocking.
Environment
Tecnai TF20
Resolution
These are possible resolutions for the issue:
  • Explain to customer.
Cause
These are possible causes for the issue:
  • Normal behavior.
Details
It is a normal behavior to see CRT axis, CAS axis and DOC temperature raise during dock and undock CAS. There is a test to verify if the temperature raise in AL is within spec, following DOC. 4022 190 90179 chapter 5. It is important to meet the precondition before the temperature test. It may take 15 min to meet the precondition temperature especially DOC as it’s holding the CAS.
CAS axis precondition temperature are the highest as it take longer time to cool compare to CRT. Yet CAS and CRT have the same spec. This imply CAS has less buffer to cater to temperature raise during CAS exchange.
Care must be taken to ensure before docking, CAS is cool to LN2 temperature inside the capsule. When LN2 is not boiling, this is an indication CAS is cool close to LN2 temperature.
clipboard_e249a89fd6236a00bbcf7ec745e5c8693.png
clipboard_e98314117fb2b22826b781fa0f353e2e9.png
clipboard_ecd444098a8823ba1e01b361a4caf1907.png
  Pre-Cond Spec Delta
DOC 108 118 10
CRT 108 153 45
CAS 118 153 35
  • Was this article helpful?