ODP Turns Off
Issue
ODP turns off after V3 open
Environment
Tecnai G2 Spirit BioTWIN
Resolution
These are possible resolutions for the issue:
- Adjust P1 trip voltage
Cause
These are possible causes for the issue:
- P1 trip level voltage too low
Details
There is error in the vacuum dialogue after vacuum turn on. ‘PLCB detected an interlock. As a result the PLCB refuses to ‘Turn on’ element ‘ODP1’. ‘The PLCB of the Vacuum System detect that element ‘ODP1’ spontaneously ‘Turns on’. PLCB dialogue shows OD_TM_ON_REFUSED. However ODP still continues to warm up. When ODP temperature is ready, V3 opens and vacuum status changes to Column Valve ready, but after a while the ODP turns off. At that moment, the vacuum dialog shows ‘The watchdog of the vacuum system detected the following error: ‘Projection (P3) pressure too high’; PLCB dialogue shows no errors anymore.
Replace P3 gauge, top up ODP oil, check CDB fuse and jumper, replace PLCB, check ODP heater resistance, re-install SW, replace MOB and MVG all did not help.
After adjusting P1 trip voltage, problem was solved (instructions on next slide).
Attachment(s)
File | Last Modified |
---|---|
clipboard_ef10d27355bf325f2170f866a76717ba5.png | June 17, 2022 |
clipboard_e77278e93480752a6370547b70f8d8cd5.png | June 17, 2022 |
clipboard_e2ea59ed7d638e805129002abb6797e31.png | June 17, 2022 |