After Installed HAADF on Krios, OASA FW validation failed
Issue
After Installed HAADF on Krios, OASA FW validation failed.
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
- Follow WI 302210 manually update FW
Cause
These are possible causes for the issue:
- Desired version is different with current version in FW validation
Details
After installed HAADF on Krios, issue happened as below:
- OASA FW failed, follow WI 302210 to update FW manually using with "//", not to skip all good boards but keep one by one.
Note:
In this case License 4K*4K STEM acquisition not active, as a result customer can only get 2K images STEM.
Attachment(s)
File | Last Modified |
---|---|
clipboard_e94249cf56193d28fcdeaca1ca17423ba.png | June 23, 2022 |
clipboard_e4b268657d1f19cb04350167a57761c0d.png | June 23, 2022 |