Thermo Fisher Scientific
Auto CTF is not working properly
Issue
Auto CTF is not working properly
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
  • Restore default settings
Cause
These are possible causes for the issue:
  • Sherpa setting not correct
Details
Auto stigmatism and auto coma of Auto CTF in the Sherpa is not working properly. Issue fixed by restore default settings.
Notes: As this tool has Cs image corrector, so the comma correct function is not available. The coma correction has to be done in the Cs image corrector software. For the non-Cs image corrected systems, Sherpa Auto CTF uses the beam tilt to correct coma .
clipboard_e069ff4dae39a8f8dc85dd14cf7cf48b4.png clipboard_eb69a2a7fcf1119273e780a7a3713e292.png
  • Was this article helpful?