Thermo Fisher Scientific
Bruker Esprit v.1.9 NOT communicating with TEM_SERVER
Issue
Running QMap and QLine on Bruker Esprit v.1.9 Software Failures, NOT showing TEM Microscope Status & High Voltage State.
Environment
Titan G2 60-300
Resolution
These are possible resolutions for the issue:
  • Re-install Bruker Esprit v.1.9 from ISO Installer
Cause
These are possible causes for the issue:
  • Bruker Esprit v.1.9 supplement software “Quantax Communication Server” Corrupted.
Details
Documents needed:
WI-4022 190 53648 - Esprit Super-X
  • Following Chapter-3.2 - Esprit 1.9 Installation. 
    • Make sure to choose with Super-X Tecnai Osiris
    • Next also Make sure to Install SEMLink
    • And Make sure SEMLink Settings are Correct: 
      • Producer = FEI 
      • Type = Tecnai Osiris 
      • Driver = FEITEM (library integrated)
clipboard_e2ff38030df401c40aa48d96ccd1cbdbd.png
clipboard_e6a4447fb83c901dae3cfdf1adfc59beb.png
clipboard_ecbbcb4eee0728092eb11f0fc5a2365ae.png