Thermo Fisher Scientific
C3 Optimum Unit Abnormal
Issue
Image Corrector C3 optimum unit abnormal on PCTF page
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
  • Correction on Resolution g Desired value
Cause
These are possible causes for the issue:
  • Resolution g Desired value had a typo
Details
  1. FSE reported that C3 optimum unit in PCTF page is “km”.
  2. Found that After correction on the value of “Resolution g Desired value”, this unit came back to normal
clipboard_ea16e24663f979262feefc814c8346561.png
  • Was this article helpful?