Cassette would drop down from docker during loading, unloading or inventory
Issue
Cassette would drop down from docker during loading, unloading or inventory.
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
- Replace docker.
Cause
These are possible causes for the issue:
- U-bracket of front docker was bent.
Details
During verification procedure 104358, FSE found U-bracket of front docker was bent.
Action taken as below but doesn't help:
- Tried to straighten the U-bracket but there were still big movements on slots 9,10 and 11 during inventory.
- Performing Cassette Arm Park, Capsule, Docker slot 12 alignment.
Issue was solved after replacing the docker.