Thermo Fisher Scientific
Communication error ( SPI Bus )
Issue
Communication error ( SPI Bus )
Environment
Titan Cubed 80-300
Resolution
These are possible resolutions for the issue:
  • Replaced DPA
Cause
These are possible causes for the issue:
  • Defective DPA
Details
Customer loaded different FEG register and there was no beam.
There was SPI bus communication error in User Interface. Ref and Readbacks in Mono outputs were fluctuating.
Followed Doc. 106534 Monochromator Electronics Troubleshooting FEG G1 TS, Replaced CPB2 -1051379 , SDC2- 1051365. Didn’t help.
Readbacks on Mono still fluctuating & SPI bus error still appears with mono normalization.
Replaced DPA2 – 1051377 and mono refs and readbacks stopped fluctuating.
Did following measurements and check voltages with HT probe on Mono gold contacts: ( CH. 7 Doc 106534 )
Y Deflection and Y Stig check, Main Deflection and X stig check, Icoil current check
All measured values matched with set values for mono electronics.
Solution : Replace DPA board
clipboard_ebb14044194557500e0ed096b4881d5d1.png
clipboard_ee5a0521679b20da813c185d3d24e08b0.png