Comparing EPU 2.8 to 2.7, 2.8 file size is 4-8X larger.
Issue
Comparing EPU 2.8 to 2.7, 2.8 file size is 4-8X larger.
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
- Upgrade EPU to the version of 2.8.1.
Cause
These are possible causes for the issue:
- Incorrect output format with LZW compression.
Details
Even though the same parameters were used in the EPU data acquisition, the TIFF format file size is >8X in EPU 2.8. There were 3 major SW bugs in EPU 2.8. One of them is the LZW compression.
The solution for this issue is to upgrade the EPU software to 2.8.1 which was released on 24 Aug 2020.
EPU 2.7
EPU 2.8
Attachment(s)
File | Last Modified |
---|---|
clipboard_e2e7d0212221aab8ffe14713e5fab0e26.png | June 20, 2022 |
clipboard_ed839408d23cb9774b02339a20ca8f533.png | June 20, 2022 |