Thermo Fisher Scientific
Customer Dongle TrueImage is missing
Issue
TrueImage Feature Application of “Automation ExperimentsOCX found missing and unable to work.
Environment
Themis Z (3.1)
Resolution
These are possible resolutions for the issue:
  • Download New Dongle Update File from “BOX – Customer Dongle + Licenses” Folder, and Perform V2C-way Dongle upgrade Procedures. Next Step, is to Re-install TEM Software to Enabling Configurator Checkbox of “TrueImage 5470/10”.
Cause
These are possible causes for the issue:
  • TrueImage” is NOT Enabled at Customer Dongle Licenses.
Details
Documents needed:
WI-4022 190 53598 - Customer Dongle and Software Protection
  • Referring to Chapter-6.3 - Updating the Sentinel EMS USB customer dongle.
How to Download Dongle Upgrade File from BOX: 
  • Login to BOX. 
  • Go to “Customer Dongle + Licenses” Folder, then search by serial number (e.g., D3808 or 9930146). 
  • Download the attached V2C file – “D3808 – 9930146 – add 54 70101 – 3aug2020.V2C”.
V2C-way Dongle Upgrade short Procedure: 
clipboard_e34d3f98bc704351ddde05cdeac25bbce.png
clipboard_eb33f3d040e73a3a55410c72fc9ff071d.png
Reminder: 
  • After Dongle upgrade and confirmed “TrueImage” License had been found OK with “License Reader”. 
  • Next step is to Re-install Full software ISO installer package again to make “Configurator” Checkbox Enabling the “Automation Experiments” Application.
clipboard_e67a52f850230bb304234bfdd8627d954.png 
How to perform V2C File to Upgrade Dongle: 
  • Go to Update/Attach Options. 
  • Go to Browse for where you save these V2C Upgrade File, which download from BOX. 
  • Next, Apply File.
clipboard_e58a4c081a7e2e16bdb9dcf1dc13448bc.png
  • Was this article helpful?