Thermo Fisher Scientific
Customer dongle update fails
Issue
customer dongle update fails with error message “another update must be installed first”
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
  • re-apply previous dongle update file first, then apply new update file
Cause
These are possible causes for the issue:
  • previous dongle update was not fully applied
Details
Original problem was that the customer dongle was missing the license for “TEM AutoCTF PhasePlate activation”. This was reported to Orderdesk Acht (NLEIN.Order-desk-Acc@thermofisher.com) who confirmed that the item is missing and provided a new customer dongle update file.
The update file on the system failed with error “another update must be installed first”.
We logged into the Sentinel server and compared the contents of the previous update with the current customer dongle status on the system, then found that several items of the previous update file were not enabled.
We asked to run the previous customer dongle update first, then the “new” update, then the problem was solved.
Error message during customer dongle update (“new” update file)
clipboard_ee629056cb0999ba5ec210398364daaac.png
Capture.JPG
  • Was this article helpful?