Thermo Fisher Scientific
EPU “Error when acquiring image! Acquisition label must be unique”
Issue
When Acquiring an image in EPU in fraction mode with Falcon3, EPU returns : “Error when acquiring image! Acquisition label must be unique”
Environment
Titan Krios
Resolution
These are possible resolutions for the issue:
  • Reset CMTS
Cause
These are possible causes for the issue:
  • CMTS did not transferred images to storage server
Details
Goto Falcon service tool\storage\ => Found : Storage number of stored dataset = 711, not 0 => images not transfered to the server => Push the "initialized storage” button, then “refresh” => number of stored dataset = 0
clipboard_efa3c238abeab3867421e313cfd48e7b1.png
clipboard_e483eb0cb88e9040a707695a00bcf89c1.png
  • Was this article helpful?