EPU Atlas not complete, bottom RHS of atlas is missing one tile
Issue
EPU Atlas not complete, bottom RHS of atlas is missing one tile.
Environment
Krios G3
Resolution
These are possible resolutions for the issue:
- Use higher mag.
Cause
These are possible causes for the issue:
- EPU setting for K3 not optimize. Distortion at lower mag.
Details
EPU Atlas not complete, bottom RHS of atlas is missing one tile, while there is no problem when using Ceta and Falcon.
In the EPU manual DOC. 1025707 chapter 10.1 Troubleshooting Atlas, One of the solution for this issue is to use higher mag because there is inheritance distortion in the image at lower mag.
Notes: For this kind of issue other suggestions should be taken into consideration as suggested in the WI-1025707. It could be a combination of causes.
Incomplete Atlas in K3 ~LM 100X
Attachment(s)
File | Last Modified |
---|---|
clipboard_e92aa9b42a9d24e217782562bd18c0840.png | June 20, 2022 |
clipboard_ebc90e626af01e597a6196e23ab7fbc46.png | June 20, 2022 |