Thermo Fisher Scientific
EPU and Tomography Software unable to Open and Error
Issue
EPU and Tomography Software unable to Open, and Error Messages:Problem encountered while checking the license for EPU Exception from HRESULT: 0x800000BF”.
Environment
Titan Krios G3
Resolution
These are possible resolutions for the issue:
  • Perform Bergson Runtime software upgrade to v.2.13.1937. 
    • Runtime can also be found in Titan v.2.11.x or higher ISO Installer Packages.
Cause
These are possible causes for the issue:
  • Caused by, the Bergson B’Protected Runtime Software too old.
Details
Troubleshooting Details under Remote TeamViewer Session: 
  • Go to C:\Program Files (x86)\Common Files\Bergson Shared\Bprotected\BProtected.exe. 
  • Both EPU and Tomography Software are being Activated in Bergson B’Protected. 
    • EPU and Tomography TEM are Activated
  • By looking at the “About B’Protected”. 
    • Found this Runtime version is too old. 
  • Therefore, Perform upgrade to v.2.13.1937. 
    • After upgrade EPU and Tomography can be Opened Normally.
clipboard_e9ed260a59905cab49a3263888f45ad0a.png
clipboard_e381137ba3f0dee932d1aea95049d0b89.png
clipboard_eacb10b1dc1571c823d1a2d176afd4518.png