EPU not start up after upgraded
Issue
EPU can't start up after upgrade , Versions 2.4 is working normal and version 2.5. 2.6 and 2.6.1 with error.
Environment
Titan Halo
Resolution
These are possible resolutions for the issue:
- Delete roaming folder in supervisor account
Cause
These are possible causes for the issue:
- Roaming folder in supervisor account
Details
- Reinstall the TEM Server , issue not fixed.
- Rename in C:\ProgramData\Thermo Scientific EPU\Configuration: file : "Omp.config" remame to "Omp.config.BUG", and restart EPU. Not working.
- Uninstall EPU in supervisor. Go to directory users\app data\roaming\ EPU, rename MC settings file . Not help.
- Check C++ distribute version and dump the EPU process by task manager, not work.
- Check install and start in service account, worked at service account. Login as supervisor, not work.
- Delete content of roaming folder in supervisor, issue gone.