EnergyFilterMonitor Offline
Issue
EnergyFilterMonitoris Offline
Environment
Titan Krios
Resolution
These are possible resolutions for the issue:
- Dongle update
Cause
These are possible causes for the issue:
- No selectris in customer dongle
Details
During selectrisinstallation it is not possible to get the EnergyFilterMonitorprogram online.
In the bottom it says connection succeeded but it remains offline ( see attached image ).
Solution : The selectris option needs to be enabled in the customer dongle ( it is not by default enabled in the service dongle ) to make it work.
Attachment(s)
File | Last Modified |
---|---|
clipboard_e171d87be16f0ea346f71ad62dbbc4f8c.png | June 19, 2022 |