Thermo Fisher Scientific
FEG shuts down randomly
Issue
FEG shuts down randomly without error message.
Environment
Titan Cubed G2 60-300
Resolution
These are possible resolutions for the issue:
  • Repair or replace the damaged cables.
Cause
These are possible causes for the issue:
  • FIB/H cables were damaged due to arcing.
Details
Based on the log file, there are a lot of CPBF communications errors being lost and restored.
While FSE was replacing CPB board, he found some damage on the cable between FIB/H X104 and MWB X10 and he also found burnt marks on FIB/H X11 cable.
Issue seems to be solved after FSE repaired the cable on site but it is advisable to replace the damaged cables if possible.
clipboard_ee73c5604e8872356560d1dff6bc5b037.png
clipboard_e72797d2a187e80e6a22a73059ee3ac90.png
clipboard_e08de9ea9730efbc15f2fedba942d5180.png
clipboard_eb09ffacc953be8f8aeec0d63a5294c06.png
  • Was this article helpful?