Flu screen inserting error and crash flu screen viewer
Issue
when insert the flu screen, flu screen viewer crash,Screen lift button doesn't work with error message:"HRESULT E_FAIL has been returned from a call to a COM component"
Environment
Titan 80-300
Resolution
These are possible resolutions for the issue:
- replace Flu screen assembly
Cause
These are possible causes for the issue:
- Flu screen assembly defective
Details
Flu screen viewer crashes with an error during each screen inserting. Screen lift button on the hand panel doesn't work as well.
Action:
Replacing the UIOB flu and running a firmware update.---->No help.
TAD test of UIOB flu almost all pass except self test due to firmware not allow
All other critical test pass.
Checked the sensors on the lifting mechanism---> both working when manual insert/retract.
Solution: Replacing the entire flu screen assembly , issue fixed.
Error message flu screen inserting
Self test not allow
Attachment(s)
File | Last Modified |
---|---|
clipboard_ec9e9bbc3a85ce8ef517f21702ccdfa77.png | June 18, 2022 |
clipboard_ee342a9225adf61d6cf4f6a6e06de2bbb.png | June 18, 2022 |