Fluorescent screen insert/retract is much slower than normal
Issue
Fluorescent screen insert/retract is much slower than normal.
Environment
Krios
Resolution
These are possible resolutions for the issue:
- Replace fluscreen drive assy.
Cause
These are possible causes for the issue:
- Cylinder end of life.
Details
Fluorescent screen inserting and retracting is much slower than normal.
Check no mechanical obstruction.
Check air pressure is sufficient visually.
The cylinder is not moving smoothly.
PS: Need to order fluscreen rotation assembly (PN 1131759) instead of cylinder, cost is less as a FRU.
Note: In case of large attachments (for example video), please make a note here to go to the TA.
Attachment(s)
File | Last Modified |
---|---|
clipboard_e445da595ddc603abf9b1ee16607ce4d3.png | June 19, 2022 |
clipboard_ed09bf83cea73ef10524bf37dc5fd0c04.png | June 19, 2022 |