Fluscreen is not raising to correct position
Issue
Fluscreen is only raising to half of the actual height each time screen lift is pressed.
Environment
Titan G2 60-300
Resolution
These are possible resolutions for the issue:
- Replace SPB.
Cause
These are possible causes for the issue:
- Defective SPB.
Details
This system has the motorized screen lift and not the pneumatic one.
Each time the screen lift is pressed, the screen can be raised but not to the correct position.
The motor was replaced but did not solve the issue. The issue was solved after replacing the SPB- 4022 268 00847.