Thermo Fisher Scientific
HASP Service Dongle key not Identified~Detected
Issue
LPT-Port (HASP type) Service Dongle Key not Identified and not Detected, unable to Login TEM_UI under customer service account.
Environment
Krios G3i (3.1)
Resolution
These are possible resolutions for the issue:
  • Make sure the Hardware Tree in “Device Manager” can Identify with Ports (COM & LPT) having “Printer Port (LPT1)” being Enabled
  • Re-Plug and Re-Slot this LPT1 Cable Extension on Motherboard located with LPT1 Header-Pins.
Cause
These are possible causes for the issue:
  • MPC Motherboard “LPT1” Cable Extension Connector had found Loosen.
Details
Documents needed:
WI-103405 - Microscope PC
  • Check Device Manager
    • Make sure Printer Port (LPT1) is being Enabled
  • Following Chapter-4.3 - IMBA PC (Win7). 
    • Make sure this is a Win7 Base MPC. 
    • Referring to Chapter-4.3.3 - Mechanical Settings Specification, to find where this LPT1 Head-Pins to be located. 
    • Problem fixed by Re-Plug and Re-Slot this Cable Extension.
clipboard_e33fb141dad7986597c97332417be2749.png
clipboard_e2999e160e3fae5d305758bd4ae3614ec.png
00499.png
  • Was this article helpful?