Thermo Fisher Scientific
HT Can’t Raise to 300KV
Issue
HT can’t raise to 300 kV
Environment
Titan Krios G3
Resolution
These are possible resolutions for the issue:
  • Replace HT tank
Cause
These are possible causes for the issue:
  • HT tank defect
Details
Power cycled HT tank, does not help.
Measured bleeder resistor (HT tank test pins X241 vs. X242).
Measured measuring resistor (HT tank test pins X104 vs. X106).
Bleeder and measuring resistor for 300 kV tank should give 1.3V.
From HT 237 kV, the voltages measured on the bleeding and measuring resistors becomes unstable. At the same time, the HT level becomes unstable.
HT tank is deemed defective (problem inside HT tank).
Problem solved after replace tank.
Capture.JPG
clipboard_e7a02ae73af3466c166da6a5c0ed6f37f.png
HT raises to 280 kV, then drops to 60 kV and then jumps between 120~280 kV.
clipboard_e4b326ecae84b1dd89790a482d270efd5.png