HT Can’t Start,Gun dialog shows “External interlock”
Issue
HT cannot start,Gun dialog shows “External interlock”
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
- Replace VGIB
Cause
These are possible causes for the issue:
- Defected VGIB cause external vacuum interlock
Details
- FEG is normal, but HT cannot start.
- Almost all HT TAD tests failed on “HT on conditions” or “check HT and AUX are switched off”
- Gun dialog shows “External interlock”
- PILZ position was verified
- K1 on IETM is the vacuum interlock relay, follow this path and found that VCBT is working properly, but VGIB is not generating output voltage back to IETM
- Replace VGIB and the issue was fixed
Attachment(s)
File | Last Modified |
---|---|
clipboard_ea1c5b2ebf4389f79c9ac88544c093ad8.png | June 17, 2022 |
clipboard_efb2fa1b4e74b0945b9b13e4c84a1a8d2.png | June 17, 2022 |