Thermo Fisher Scientific
HT Cannot reach 300kV
Issue
HT cannot reach 300kV
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
  • Replace HT Tank
Cause
These are possible causes for the issue:
  • HT Tank is defective
Details
  1. High tension cannot reach maximum value, when it reaches 296kV it will drop to around 100kV, then it will rise again then fluctuate between 220kV and 270kV, and it will be stable around 200kV
  2. Inspected HT cable and visual check is passed.
  3. HT Generator Test: the result is the same as the symptom of this issue 4HT
  4. Generator Test with flange:
    • First time it is identical to the symptom of this issue
    • From the 2nd and 6th time, the test passed
    • Last time it failed again
  5. HTPOB replaced
  6. Double check the HT cables’ isolation
  7. Replaced HT Tank and the issue was fixed
clipboard_e38a68542cfe3a8f76ba99e93bee14385.png