Hand Panel not responding after MPC reboot
Issue
After MPC is rebooted RHP will not connect
Environment
Metrios G2
Resolution
These are possible resolutions for the issue:
- Install wired mouse and keyboard.
Cause
These are possible causes for the issue:
- Customer installed wireless mouse and keyboard.
Details
RHP will not connect after rebooting MPC. Work around is to disconnect LHP then reconnect LHP . This establishes connection to RHP. All TAD tests for the RHP fail when connected ( working ) or disconnected ( not working ). We have replaced all RHP hardware. Hand panels, cables, interconnect boxes, MPC, were replaced per GTS and problem still persists. SW upgrade to latest revision of SW didn't resolve issue either.