LicenseManagementService not running
Issue
Unable to activate EPU and Tomo.
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
- Uninstall and then reinstall LicenseManagementService. After that, reinstall Bergson version 2.13.
Cause
These are possible causes for the issue:
- LicenseManagementService was not running and could not be started.
Details
Uninstall and then reinstall ‘LicenceManagementDeveloper64' and 'LicenceManagementRuntime64’ from SW 2.14 Applications.iso because for unknown reason, we could not find the ‘LicenceManagementDeveloper64' and 'LicenceManagementRuntime64’ in SW 2.15.3 Applications.iso.
After that, the LicenseManagementService working and then we register the products again in PkLicensing.
However we encountered a different error while opening EPU and Tomo, after reinstalling Bergson version 2.13, issue fixed.
Error while running BProtected.exe
Error while running PKLicensing.exe
Error after opening EPU
Attachment(s)
File | Last Modified |
---|---|
clipboard_ea4ebf6522d370ac0e1d297cde5766cb0.png | June 20, 2022 |
clipboard_e2922a13cfb2028510ff817ae858dc573.png | June 20, 2022 |
clipboard_e266c108886c21cacd408c4c62656976f.png | June 20, 2022 |