Mono instability in Y direction
Issue
Mono instability in Y direction.
Environment
Titan 80-300
Resolution
These are possible resolutions for the issue:
- Replaced 3 boards of GMS, DPA and SDCA.
Cause
These are possible causes for the issue:
- Boards defective.
Details
Emitter was replaced 2 months ago, at that time FEG can’t be started, it had communication problem, replacing GMS, SES, CPB boards fixed it.
Now FEG emission current = 589 uA, before it was about 400 uA.
To determine if the instability source is HT or FEG or charging related:
- HT emission current = 0 uA.
- Back focal plane check per WI-300259 at 300 and 80 kV.
- HT AC stability test per WI-103190 in service and normal mode by HCA X‐100 jumper, normal mode OK (< 500 mVpp).
- HFP test per WI-105031, Ibleeder and Imeasuring-resistor and Cascade OK.
- FREF test from TAD, OK.
- Follow WI-106115 “Mono instability TFS”, and goes to HT conditioning, not favorable.
- Source of instability confirmed to be in FEG (WI-306046, WI-106115).
Attachment(s)
File | Last Modified |
---|---|
clipboard_ea9ce9da4d92741bb086cb7ccb63f5cdf.png | June 20, 2022 |
clipboard_e5f1c13881649cede76f8a22859cd30a9.png | June 20, 2022 |
clipboard_ecbb938bdd9ded4eae600682516778d0c.png | June 20, 2022 |