OSD cannot start up with error 404
Issue
OSD cannot start up with error 404
Environment
Krios G4
Resolution
These are possible resolutions for the issue:
- From Task Manager restart Apache service
Cause
These are possible causes for the issue:
- Apache service is not working
Details
- OSD power cycle didn’t work
- Restart TEM server didn’t work
- Restart MPC didn’t work
- Root cause is Apache service is not working
- Solution: From Task manager restart Apache service
Note: We should stop apache service first and start it again
Attachment(s)
File | Last Modified |
---|---|
clipboard_e63b038413cd6f3e54598604d5db5d38b.png | June 18, 2022 |