Projection chamber vacuum leak
Issue
when HAADF inserted projection chamber can only pump to 54 logs
Environment
Titan G2 60-300
Resolution
These are possible resolutions for the issue:
- Replace HAADF
Cause
These are possible causes for the issue:
- HAADF caused the leak
Details
Projection chamber vacuum leak to air after HAADF retracted, CCGp crashed to be disabled
When HAADF detector inserted, projection chamber vac can pump down to 54 logs
After removal of HAADF detector, vac can reach 27logs with dummy flange So have HAADF replaced and fixed the issue
Notes: Technically the Oring between column and HAADF should be replaced first before exchanging HAADF. But in this case HAADF was ordered to fix this issue due to other reasons .