Thermo Fisher Scientific
Sample jumps as stage is tilted
Issue
Sample jumps at different positions when stage is tilted .
Environment
Krios G3i
Resolution
These are possible resolutions for the issue:
  • Replace the holder
Cause
These are possible causes for the issue:
  • Holder tip loosen
Details
  1. Fix x donator. No improvement for sample jumping.
  2. Adjust the tightness of y donator. No improvement for sample jumping
  3. Check behavior when krios is warm. Issue disappears when scope is warmed up and returns back when system cool down.
  4. Check the stage securing bolts behind the half moons. Very tighten.
  5. Tap the stage. it slightly moves in the tap direction but stops Immediately. Floating rod is stable.
  6. Test with another holder, Sample doesn't jump at all. Check the holder and found the tip on the holder is loosen.
  • Was this article helpful?