Thermo Fisher Scientific
Spontaneous system restart after touching LCD screen
Issue
Vitrobot Spontaneously restarted after touching LCD screen when it is idle.
Environment
Krios G4 (4.0)
Resolution
These are possible resolutions for the issue:
  • Adjust the supply to PC to 5 volts.
Cause
These are possible causes for the issue:
  • 5 V supply for PC is not stable.
Details
When Vitrobot is idle, the screen is normally black, if customer touch the screen, the Vitrobot restart sometimes.
Action taken as below:
  1. PC104 replaced already → issue not fixed.
  2. It can be LCD problem sending wrong signal to system. So check the LCD's cabling and recalibrate LCD → no help.
  3. We found the 5 volts for PC104 was actually 4.43 V, which was too low, adjust it to 4.78 max by the potential meter (refer to the second image below) and then we cut the tie wrap of PC104 and manage to get 5.18 V → issue fixed.
clipboard_e66dbc49c2cb276dc5ea0cdfd6cbb5db0.png
clipboard_ecab0d2a22fbe651beee45dabf99afb90.png