Tomography v.5.2.0 - Tilt Series showing Stage Jumps~Shifts from Image to Image
Issue
Tomography v.5.2.0 – Tilt-Series showing Stage Jumps~Shifts from Image to Image.
Environment
Krios G3i (3.1)
Resolution
These are possible resolutions for the issue:
- From the “Corrections” Section at Title-Bar for parameter settings of “Tracking – Switch Angle(°)” Option.
- Change from 10 degrees to 80 degrees.
Cause
These are possible causes for the issue:
- Switch angle setting too small.
Details
How to Modify “Tracking – Switch Angle(°)”:
- Go to the “Corrections” Section at Title-Bar.
- At the input Box of “Switch Angle” must be Gray-out unable to access.
- Gray-out to be protection for sudden modification.
- Tick the Checkbox of “Track Before Acquisition”.
- This will Free-out the Access for new inputs.
- Now to modify these “Switch Angle”.
- Change from 10 Degrees to 80 Degrees.
- Enable for Stage to Actively perform Tilt-Series Corrections up to 80 Degrees.
Attachment(s)
File | Last Modified |
---|---|
clipboard_e733661247fce100a22c19b38abac6841.png | June 20, 2022 |